siRNA Details
| Gene Symbol: CCDC92B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_017025489.1 | XP_016880978.1 | 3 | 219 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

CCDC92B| Gene Symbol: CCDC92B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_017025489.1 | XP_016880978.1 | 3 | 219 | ||