siRNA Details
Gene Symbol: GRAMD1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017018036.1 | XP_016873525.1 | 1 | 557 | ||
XM_017018037.1 | XP_016873526.1 | 2 | 856 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: GRAMD1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017018036.1 | XP_016873525.1 | 1 | 557 | ||
XM_017018037.1 | XP_016873526.1 | 2 | 856 |