siRNA Details
| Gene Symbol: GRAMD1B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_017018034.1 | XP_016873523.1 | 2 | 294 | ||
| XM_017018035.1 | XP_016873524.1 | 2 | 294 | |||
| XM_017018036.1 | XP_016873525.1 | 1 | 761 | |||
| XM_017018037.1 | XP_016873526.1 | 2 | 1060 | |||
| XM_017018038.1 | XP_016873527.1 | 2 | 294 | |||

GRAMD1B
Genomic Map