siRNA Details
| Gene Symbol: TMEM210 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001282477.1 | NP_001269406.1 | 3, 4 | 264 | ||
| GenBank mRNA | AI208887.1 | - | 1, 2 | 248 | ||
| DB459119.1 | - | 3, 4 | 322 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

TMEM210| Gene Symbol: TMEM210 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001282477.1 | NP_001269406.1 | 3, 4 | 264 | ||
| GenBank mRNA | AI208887.1 | - | 1, 2 | 248 | ||
| DB459119.1 | - | 3, 4 | 322 | |||