siRNA Details
| Gene Symbol: MID1IP1-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_046706.1 | 1 | 118 | |||
| GenBank mRNA | DY655415.1 | - | 1 | 28 | ||
| Gene Symbol: MID1IP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021242.5 | NP_067065.1 | 2 | 1284 | ||

MID1IP1-AS1
Genomic Map