siRNA Details
| Gene Symbol: MID1IP1-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_046706.1 | 2 | 189 | |||
| GenBank mRNA | DY655415.1 | - | 2 | 99 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

MID1IP1-AS1| Gene Symbol: MID1IP1-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_046706.1 | 2 | 189 | |||
| GenBank mRNA | DY655415.1 | - | 2 | 99 | ||