siRNA Details
| Gene Symbol: ASPSCR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK090682.1 | - | 1 | 848 | ||
| AK126967.1 | - | 1 | 3143 | |||
| AK127159.1 | - | 1 | 55 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

ASPSCR1| Gene Symbol: ASPSCR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK090682.1 | - | 1 | 848 | ||
| AK126967.1 | - | 1 | 3143 | |||
| AK127159.1 | - | 1 | 55 | |||