siRNA Details
Gene Symbol: RAD54B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001205262.2 | NP_001192191.1 | 4 | 934 | ||
GenBank mRNA | BC033710.2 | - | 3 | 819 | ||
BF766767.1 | - | 1 | 39 |
Gene Symbol: FSBP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256141.1 | NP_001243070.1 | 2 | 957 | ||
GenBank mRNA | AF007866.1 | - | 2 | 909 |
