siRNA Details
| Gene Symbol: CUEDC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001271875.1 | NP_001258804.1 | 11 | 2834 | ||
| NM_001292025.1 | NP_001278954.1 | 11 | 3139 | |||
| XM_006721900.2 | XP_006721963.1 | 11 | 3449 | |||
| XM_011524812.2 | XP_011523114.1 | 11 | 2222 | |||
| XM_017024640.1 | XP_016880129.1 | 10 | 2720 | |||
| XM_017024641.1 | XP_016880130.1 | 12 | 2851 | |||
| GenBank mRNA | CR627470.1 | - | 5 | 2445 | ||

CUEDC1
Genomic Map