siRNA Details
| Gene Symbol: ASTN2-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_033973.1 | 11, 12 | 1463 | |||
| GenBank mRNA | AK057447.1 | - | 11, 12 | 1463 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

ASTN2-AS1| Gene Symbol: ASTN2-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_033973.1 | 11, 12 | 1463 | |||
| GenBank mRNA | AK057447.1 | - | 11, 12 | 1463 | ||