siRNA Details
| Gene Symbol: HPN-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_024561.1 | 2 | 125 | |||
| NR_024562.1 | 3 | 341 | ||||
| GenBank mRNA | AK124779.1 | - | 2 | 125 | ||
| AK295317.1 | - | 3 | 341 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

HPN-AS1| Gene Symbol: HPN-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_024561.1 | 2 | 125 | |||
| NR_024562.1 | 3 | 341 | ||||
| GenBank mRNA | AK124779.1 | - | 2 | 125 | ||
| AK295317.1 | - | 3 | 341 | |||