siRNA Details
| Gene Symbol: LOC102724691 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110603.1 | 3 | 148 | |||
| GenBank mRNA | BX109606.1 | - | 3 | 148 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC102724691| Gene Symbol: LOC102724691 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110603.1 | 3 | 148 | |||
| GenBank mRNA | BX109606.1 | - | 3 | 148 | ||