siRNA Details
| Gene Symbol: LINC00643 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015358.2 | 4 | 615 | |||
| GenBank mRNA | AK096461.1 | - | 1 | 137 | ||
| AK311468.1 | - | 4 | 819 | |||
| BX248751.1 | - | 4 | 493 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC00643| Gene Symbol: LINC00643 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015358.2 | 4 | 615 | |||
| GenBank mRNA | AK096461.1 | - | 1 | 137 | ||
| AK311468.1 | - | 4 | 819 | |||
| BX248751.1 | - | 4 | 493 | |||