siRNA Details
| Gene Symbol: MFAP3L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_017008865.1 | XP_016864354.1 | 1 | 1367 | ||
| XM_017008869.1 | XP_016864358.1 | 1 | 1367 | |||
| XM_017008870.1 | XP_016864359.1 | 1 | 1367 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

MFAP3L| Gene Symbol: MFAP3L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_017008865.1 | XP_016864354.1 | 1 | 1367 | ||
| XM_017008869.1 | XP_016864358.1 | 1 | 1367 | |||
| XM_017008870.1 | XP_016864359.1 | 1 | 1367 | |||