siRNA Details
| Gene Symbol: SCOC-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_033939.1 | 4 | 377 | |||
| GenBank mRNA | AK095554.1 | - | 4 | 345 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

SCOC-AS1| Gene Symbol: SCOC-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_033939.1 | 4 | 377 | |||
| GenBank mRNA | AK095554.1 | - | 4 | 345 | ||