siRNA Details
| Gene Symbol: FBRS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105079.2 | NP_001098549.2 | 5 | 722 | ||
| XM_011545916.2 | XP_011544218.1 | 5 | 2099 | |||
| XM_011545917.2 | XP_011544219.1 | 4 | 2069 | |||
| XM_011545919.2 | XP_011544221.1 | 3 | 2032 | |||
| GenBank mRNA | AK096322.1 | - | 3 | 253 | ||
| DB082271.1 | - | 5 | 319 | |||
| DB093195.1 | - | 3 | 509 | |||

FBRS
Genomic Map