siRNA Details
| Gene Symbol: LOC101929095 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_125911.1 | 5 | 336 | |||
| GenBank mRNA | DB497821.1 | - | 5 | 336 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101929095| Gene Symbol: LOC101929095 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_125911.1 | 5 | 336 | |||
| GenBank mRNA | DB497821.1 | - | 5 | 336 | ||