siRNA Details
| Gene Symbol: KLHDC4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK092895.1 | - | 1 | 429 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

KLHDC4| Gene Symbol: KLHDC4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK092895.1 | - | 1 | 429 | ||