siRNA Details
| Gene Symbol: HBCBP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AF529371.1 | - | 1 | 326 | ||
| Gene Symbol: GPR182 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_011537827.2 | XP_011536129.1 | 3 | 2040 | ||

HBCBP
Genomic Map