siRNA Details
| Gene Symbol: LOC105375650 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_132986.1 | 1 | 164 | |||
| GenBank mRNA | BC128222.1 | - | 1 | 603 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC105375650| Gene Symbol: LOC105375650 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_132986.1 | 1 | 164 | |||
| GenBank mRNA | BC128222.1 | - | 1 | 603 | ||