siRNA Details
| Gene Symbol: LOC105375650 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_132987.1 | 1 | 105 | |||
| GenBank mRNA | BC128222.1 | - | 2 | 714 | ||
| Gene Symbol: LOC286149 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK093190.1 | - | 1 | 105 | ||

LOC105375650
Genomic Map