siRNA Details
Gene Symbol: AATBC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026961.1 | 1 | 3153 | |||
GenBank mRNA | AK093410.1 | - | 2 | 168 | ||
AK128598.1 | - | 1 | 3154 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: AATBC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026961.1 | 1 | 3153 | |||
GenBank mRNA | AK093410.1 | - | 2 | 168 | ||
AK128598.1 | - | 1 | 3154 |