siRNA Details
| Gene Symbol: ZXDC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK093796.1 | - | 1 | 1065 | ||
| AY358102.1 | - | 1 | 246 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

ZXDC| Gene Symbol: ZXDC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK093796.1 | - | 1 | 1065 | ||
| AY358102.1 | - | 1 | 246 | |||