siRNA Details
| Gene Symbol: TMEM212 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001164436.1 | NP_001157908.1 | 1, 2 | 190 | ||
| XM_011512817.1 | XP_011511119.1 | 1, 2 | 190 | |||
| GenBank mRNA | AK026825.1 | - | 1, 2 | 170 | ||
| DB199311.1 | - | 1, 2 | 190 | |||

TMEM212
Genomic Map