siRNA Details
| Gene Symbol: LOC102724050 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_120488.1 | 3 | 269 | |||
| GenBank mRNA | AI809671.1 | - | 1 | 248 | ||
| DA757660.1 | - | 3 | 150 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC102724050| Gene Symbol: LOC102724050 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_120488.1 | 3 | 269 | |||
| GenBank mRNA | AI809671.1 | - | 1 | 248 | ||
| DA757660.1 | - | 3 | 150 | |||