siRNA Details
| Gene Symbol: LOC102724050 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | DA757660.1 | - | 3, 4 | 191 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC102724050| Gene Symbol: LOC102724050 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | DA757660.1 | - | 3, 4 | 191 | ||