siRNA Details
| Gene Symbol: LOC102724050 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_120487.1 | 3 | 367 | |||
| NR_120488.1 | 2 | 220 | ||||
| GenBank mRNA | AI809671.1 | - | 2 | 297 | ||
| DA746386.1 | - | 3 | 369 | |||
| DA757660.1 | - | 2 | 101 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC102724050| Gene Symbol: LOC102724050 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_120487.1 | 3 | 367 | |||
| NR_120488.1 | 2 | 220 | ||||
| GenBank mRNA | AI809671.1 | - | 2 | 297 | ||
| DA746386.1 | - | 3 | 369 | |||
| DA757660.1 | - | 2 | 101 | |||