siRNA Details
Gene Symbol: C8orf89 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001243237.1 | NP_001230166.1 | 1 | 69 | ||
XM_011517434.2 | XP_011515736.1 | 2 | 171 | |||
XM_011517435.2 | XP_011515737.1 | 2 | 208 | |||
GenBank mRNA | BC150551.1 | - | 1 | 69 | ||
BC150552.1 | - | 1 | 70 |
