siRNA Details
| Gene Symbol: C8orf89 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001243237.1 | NP_001230166.1 | 4 | 408 | ||
| XM_011517434.2 | XP_011515736.1 | 5 | 510 | |||
| XM_011517435.2 | XP_011515737.1 | 5 | 547 | |||
| GenBank mRNA | BC150551.1 | - | 4 | 340 | ||
| BC150552.1 | - | 3 | 353 | |||
| DT932667.1 | - | 4 | 292 | |||

C8orf89
Genomic Map