siRNA Details
| Gene Symbol: C17orf107 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001145536.1 | NP_001139008.1 | 2 | 233 | ||
| GenBank mRNA | AK128415.1 | - | 2 | 233 | ||
| AK308530.1 | - | 2 | 468 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

C17orf107| Gene Symbol: C17orf107 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001145536.1 | NP_001139008.1 | 2 | 233 | ||
| GenBank mRNA | AK128415.1 | - | 2 | 233 | ||
| AK308530.1 | - | 2 | 468 | |||