siRNA Details
| Gene Symbol: UNQ6494 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK126399.1 | - | 1 | 1480 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

UNQ6494| Gene Symbol: UNQ6494 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK126399.1 | - | 1 | 1480 | ||