siRNA Details
| Gene Symbol: LOC100996842 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001322800.1 | NP_001309729.1 | 2 | 668 | ||
| NR_136401.1 | 2 | 211 | ||||
| XM_006721623.3 | XP_006721686.1 | 2 | 814 | |||
| GenBank mRNA | HY076064.1 | - | 2 | 212 | ||

LOC100996842
Genomic Map