siRNA Details
| Gene Symbol: GGT8P | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_003503.1 | 2 | 1513 | |||
| GenBank mRNA | AK055838.1 | - | 2 | 1513 | ||
| Gene Symbol: GGT2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_017029185.1 | XP_016884674.1 | 8 | 1813 | ||

GGT8P
Genomic Map