siRNA Details
| Gene Symbol: LOC100130468 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK126528.1 | - | 1 | 2865 | ||
| Gene Symbol: APCDD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_011525603.1 | XP_011523905.1 | 4 | 4642 | ||

LOC100130468
Genomic Map