siRNA Details
| Gene Symbol: ZNF729 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001242680.1 | NP_001229609.1 | 4 | 390 | ||
| XM_017026122.1 | XP_016881611.1 | 4 | 436 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

ZNF729| Gene Symbol: ZNF729 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001242680.1 | NP_001229609.1 | 4 | 390 | ||
| XM_017026122.1 | XP_016881611.1 | 4 | 436 | |||