siRNA Details
| Gene Symbol: C22orf46 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001142964.1 | NP_001136436.1 | 2 | 2181 | ||
| XM_011530373.2 | XP_011528675.1 | 3 | 2089 | |||
| XM_017028939.1 | XP_016884428.1 | 4 | 2220 | |||
| GenBank mRNA | AK095008.1 | - | 1 | 936 | ||
| AK131457.1 | - | 4 | 2015 | |||
| BC035403.1 | - | 2 | 2181 | |||

C22orf46
Genomic Map