siRNA Details
| Gene Symbol: LOC100130597 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK127669.1 | - | 1 | 256 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100130597| Gene Symbol: LOC100130597 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK127669.1 | - | 1 | 256 | ||