siRNA Details
| Gene Symbol: LOC100130597 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK127669.1 | - | 1 | 392 | ||
| Gene Symbol: SLC19A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_011529706.2 | XP_011528008.1 | 1 | 124 | ||

LOC100130597
Genomic Map