siRNA Details
| Gene Symbol: PRR32 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001122716.1 | NP_001116188.1 | 2 | 447 | ||
| GenBank mRNA | BG200532.1 | - | 1 | 347 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

PRR32| Gene Symbol: PRR32 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001122716.1 | NP_001116188.1 | 2 | 447 | ||
| GenBank mRNA | BG200532.1 | - | 1 | 347 | ||