siRNA Details
| Gene Symbol: TMEM35B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001195156.1 | NP_001182085.1 | 3 | 433 | ||
| GenBank mRNA | AK129824.1 | - | 3 | 411 | ||
| BC029439.1 | - | 3 | 405 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

TMEM35B| Gene Symbol: TMEM35B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001195156.1 | NP_001182085.1 | 3 | 433 | ||
| GenBank mRNA | AK129824.1 | - | 3 | 411 | ||
| BC029439.1 | - | 3 | 405 | |||