siRNA Details
| Gene Symbol: LOC100130698 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_136612.1 | 6 | 1449 | |||
| GenBank mRNA | AK128140.1 | - | 1 | 2165 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100130698| Gene Symbol: LOC100130698 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_136612.1 | 6 | 1449 | |||
| GenBank mRNA | AK128140.1 | - | 1 | 2165 | ||