siRNA Details
| Gene Symbol: HOXB-AS3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110331.1 | 3 | 298 | |||
| GenBank mRNA | CF127739.1 | - | 3 | 276 | ||
| DA741273.1 | - | 3 | 303 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

HOXB-AS3| Gene Symbol: HOXB-AS3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110331.1 | 3 | 298 | |||
| GenBank mRNA | CF127739.1 | - | 3 | 276 | ||
| DA741273.1 | - | 3 | 303 | |||