siRNA Details
| Gene Symbol: SMIM6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001162997.1 | NP_001156469.1 | 1, 2 | 229 | ||
| XM_005256903.4 | XP_005256960.1 | 2, 3 | 1707 | |||
| GenBank mRNA | AK131023.1 | - | 1, 2 | 156 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

SMIM6| Gene Symbol: SMIM6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001162997.1 | NP_001156469.1 | 1, 2 | 229 | ||
| XM_005256903.4 | XP_005256960.1 | 2, 3 | 1707 | |||
| GenBank mRNA | AK131023.1 | - | 1, 2 | 156 | ||