siRNA Details
| Gene Symbol: LINC01502 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_034016.1 | 2 | 274 | |||
| NR_034017.1 | 2 | 274 | ||||
| NR_109814.1 | 2 | 274 | ||||
| GenBank mRNA | AK098241.1 | - | 2 | 248 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC01502| Gene Symbol: LINC01502 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_034016.1 | 2 | 274 | |||
| NR_034017.1 | 2 | 274 | ||||
| NR_109814.1 | 2 | 274 | ||||
| GenBank mRNA | AK098241.1 | - | 2 | 248 | ||