siRNA Details
| Gene Symbol: LINC01502 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_034016.1 | 5, 6 | 563 | |||
| NR_034017.1 | 4, 5 | 422 | ||||
| NR_109814.1 | 3, 4 | 364 | ||||
| GenBank mRNA | AK098241.1 | - | 4, 5 | 396 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC01502| Gene Symbol: LINC01502 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_034016.1 | 5, 6 | 563 | |||
| NR_034017.1 | 4, 5 | 422 | ||||
| NR_109814.1 | 3, 4 | 364 | ||||
| GenBank mRNA | AK098241.1 | - | 4, 5 | 396 | ||