siRNA Details
| Gene Symbol: ABCB9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_011538098.2 | XP_011536400.1 | 1 | 1602 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

ABCB9| Gene Symbol: ABCB9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_011538098.2 | XP_011536400.1 | 1 | 1602 | ||