siRNA Details
| Gene Symbol: LINC01135 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_034014.1 | 2 | 231 | |||
| NR_034015.1 | 2 | 231 | ||||
| GenBank mRNA | AK055150.1 | - | 2 | 222 | ||
| AK296298.1 | - | 2 | 233 | |||
| DA202807.1 | - | 2 | 222 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC01135| Gene Symbol: LINC01135 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_034014.1 | 2 | 231 | |||
| NR_034015.1 | 2 | 231 | ||||
| GenBank mRNA | AK055150.1 | - | 2 | 222 | ||
| AK296298.1 | - | 2 | 233 | |||
| DA202807.1 | - | 2 | 222 | |||