siRNA Details
| Gene Symbol: LINC01135 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_034014.1 | 4 | 558 | |||
| NR_034015.1 | 3 | 429 | ||||
| NR_108106.1 | 2 | 256 | ||||
| GenBank mRNA | AK055150.1 | - | 4 | 549 | ||
| AK296298.1 | - | 3 | 431 | |||
| DA202807.1 | - | 4 | 549 | |||
| DA365881.1 | - | 2 | 244 | |||

LINC01135
Genomic Map