siRNA Details
| Gene Symbol: SRP14-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040059.1 | 4 | 1249 | |||
| NR_040060.1 | 3 | 966 | ||||
| GenBank mRNA | AK127696.1 | - | 3 | 964 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

SRP14-AS1| Gene Symbol: SRP14-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040059.1 | 4 | 1249 | |||
| NR_040060.1 | 3 | 966 | ||||
| GenBank mRNA | AK127696.1 | - | 3 | 964 | ||