siRNA Details
| Gene Symbol: TNRC6C-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040071.1 | 1 | 2392 | |||
| GenBank mRNA | AK094898.1 | - | 1 | 1864 | ||
| AK127699.1 | - | 1 | 433 | |||
| AY129024.1 | - | 1 | 928 | |||
| BC041417.2 | - | 1 | 2393 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

TNRC6C-AS1| Gene Symbol: TNRC6C-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040071.1 | 1 | 2392 | |||
| GenBank mRNA | AK094898.1 | - | 1 | 1864 | ||
| AK127699.1 | - | 1 | 433 | |||
| AY129024.1 | - | 1 | 928 | |||
| BC041417.2 | - | 1 | 2393 | |||